Merck
  • Synergistic inhibition of microbial sulfide production by combinations of the metabolic inhibitor nitrite and biocides.
Synergistic inhibition of microbial sulfide production by combinations of the metabolic inhibitor nitrite and biocides.
Applied and environmental microbiology (2006-09-26)
E Anne Greene, Veronique Brunelle, Gary E Jenneman, Gerrit Voordouw
ABSTRACT
MICs of six broad-spectrum biocides and two specific metabolic inhibitors and fractional inhibitory concentration indexes (FICIs) for controlling a sulfide-producing consortium were determined. Nitrite was synergistic (FICI<1) with all but one biocide due to its specific inhibition of dissimilatory sulfite reductase. Hence, combining nitrite with biocides allows more efficient and cost-effective control of sulfate-reducing bacteria.
Sigma-Aldrich
2-Bromo-2-nitro-1,3-propanediol
Supelco
Bronopol, PESTANAL®, analytical standard