Merck
  • Evidence that 4-aminobiphenyl, benzidine, and benzidine congeners produce genotoxicity through reactive oxygen species.
Evidence that 4-aminobiphenyl, benzidine, and benzidine congeners produce genotoxicity through reactive oxygen species.
Environmental and molecular mutagenesis (2007-03-21)
Patrudu Makena, King-Thom Chung
ABSTRACT
4-Aminobyphenyl (4-Ab), benzidine (Bz), and Bz congeners were evaluated for their ability to induce genotoxicity through an oxidative mechanism. The mutagenicity of these compounds was tested in the presence and absence of Aroclor 1254-induced rat S9 mix using Salmonella typhimurium tester strain TA102, which is sensitive to agents producing reactive oxygen species (ROS). In the presence of S9, 4-Ab, Bz, N-acetyl-benzidine, and 3,3-dimethoxybenzidine were strongly mutagenic in TA102, whereas, 3,3,5,5-tetra-methylbenzidine, 3,3-dimethylbenzidine (O-tolidine), and N,N-diacetylbenzidine were not mutagenic. In addition, 3,3-dichlorobenzidine and 4,4-dinitro-2-biphenylamine were directly mutagenic in TA102. Incorporation of the free radical and metal scavengers, catalase, superoxide dismutase (SOD), butylated hydroxytolune (BHT), and ethylenediamine tetraacetic acid (EDTA) reduced the mutagenic responses of 4-Ab and Bz, whereas heat-inactivated catalase and SOD had no effect. 4-Ab and Bz also induced lipid peroxidation in the presence of S9 mix as shown using the thiobarbituric acid reactive substances assay. The results of this study indicate that 4-Ab and Bz induce mutations through the induction of ROS.
Product Description
Supelco
o-Dianisidine, for spectrophotometric det. of Au, NO2-, Ce(IV), for the detection of Au, Co, Cu, SCN-, V, ≥97.0%
Sigma-Aldrich
o-Dianisidine, peroxidase substrate
Sigma-Aldrich
o-Tolidine, ≥95%
Supelco
o-Tolidine, analytical standard
Supelco
3,3′-Dichlorobenzidine, analytical standard
Sigma-Aldrich
o-Dianisidine dihydrochloride, Suitable for use in glucose determination
Sigma-Aldrich
o-Dianisidine dihydrochloride, tablet, 10 mg substrate per tablet
Sigma-Aldrich
o-Dianisidine dihydrochloride, ≥95%