Merck
  • Reagent-based, modular, tandem Michael approach for obtaining different indoline alkaloid-inspired polycyclic architectures.
Reagent-based, modular, tandem Michael approach for obtaining different indoline alkaloid-inspired polycyclic architectures.
Journal of combinatorial chemistry (2008-04-19)
Jean-Louis Brochu, Michael Prakesch, Gary D Enright, Donald M Leek, Prabhat Arya
ABSTRACT
A modular, reagent-based approach to obtain different indoline alkaloid-inspired, tetracyclic architectures is developed. With the use of TBSOTf as a Lewis acid, we report here a tandem Michael-based approach that led to the synthesis of a diastereomeric mixture of tetracyclic derivatives with two additional six-membered rings. By simply changing the Lewis acid to TMSOTf, we were able to obtain a different tetracyclic compound having additional functionalized 5- and 7-membered rings with complete stereocontrol.
Sigma-Aldrich
Trimethylsilyl trifluoromethanesulfonate, purum, ≥98.0% (T)
Sigma-Aldrich
Trimethylsilyl trifluoromethanesulfonate, 99%