Merck
  • Diastereoselective nickel-catalyzed reductive aldol cyclizations using diethylzinc as the stoichiometric reductant: scope and mechanistic insight.
Diastereoselective nickel-catalyzed reductive aldol cyclizations using diethylzinc as the stoichiometric reductant: scope and mechanistic insight.
Journal of the American Chemical Society (2008-05-17)
Pekka M Joensuu, Gordon J Murray, Euan A F Fordyce, Thomas Luebbers, Hon Wai Lam
ABSTRACT
In the presence of diethylzinc as a stoichiometric reductant, Ni(acac) 2 functions as an efficient precatalyst for the reductive aldol cyclization of alpha,beta-unsaturated carbonyl compounds tethered to a ketone electrophile through an amide or an ester linkage. The reactions are tolerant of a wide range of substitution at both alpha,beta-unsaturated carbonyl and ketone components and proceed smoothly to furnish beta-hydroxylactams and beta-hydroxylactones with generally high diastereoselectivities. A series of experiments, including deuterium-labeling studies, was carried out in an attempt to gain some insight into the possible reaction mechanisms that might be operative.
Sigma-Aldrich
Diethylzinc solution, 15 wt. % in toluene
Sigma-Aldrich
Diethylzinc, packaged for use in deposition systems
Sigma-Aldrich
Diethylzinc solution, 1.0 M in heptane
Sigma-Aldrich
Diethylzinc solution, 1.0 M in hexanes
Sigma-Aldrich
Diethylzinc, ≥52 wt. % Zn basis