Merck
Unimolecular dissociation reactions of methyl benzoate radical cation.
The journal of physical chemistry. A (2008-10-24)
Yiqun Huang, Scott Peterman, Shane E Tichy, Simon W North, David H Russell
ABSTRACT
The blackbody infrared radiation induced dissociation of methyl benzoate (C8H8O2(+*)) radical cation was investigated by using a Fourier transfer ion cyclotron resonance mass spectrometer equipped with a resistively heated (wire temperatures of 400-1070 K) wire ion guide. We observed product ion branching ratios that are strongly dependent upon wire temperature. At low temperatures (670-890 K) the major product ion C7H8 (+*) (m/z 92), which is formed by loss of CO2, and at higher temperatures (above 900 K), loss of methoxy radical ((*)OCH3) competes with loss of CO2. The energies of the various reactant ions and transition states for product ion formation were estimated by using density functional theory molecular orbital calculations, and a proposed mechanism for the dissociation chemistry of C8H8O2 (+*) involving a multistep rearrangement reaction is tested using the Master Equation formalism.
Sigma-Aldrich
Methyl benzoate, 99%
Supelco
Methyl benzoate, analytical standard
Sigma-Aldrich
Methyl benzoate, ≥98%, FCC, FG
Sigma-Aldrich
Methyl benzoate, natural, ≥98%, FCC, FG