Merck
  • Synthesis, antibacterial and antifungal activities of 3-(1,2,4-triazol-3-yl)-4-thiazolidinones.
Synthesis, antibacterial and antifungal activities of 3-(1,2,4-triazol-3-yl)-4-thiazolidinones.
Journal of enzyme inhibition and medicinal chemistry (2008-11-13)
Sumru Ozkirimli, Fusun Kazan, Yağmur Tunali
ABSTRACT
A new series of 3-(1,2,4-triazol-3-yl)-4-thiazolidinone derivatives has been synthesized by the reaction of Schiff bases of 3-amino-1,2,4-triazoles with mercaptoacetic acid and 2-mercaptopropionic acid. Their antibacterial and antifungal activities were evaluated against S. aureus, S. epidermidis, C. albicans and C. glabrata.
Sigma-Aldrich
2-Mercaptopropionic acid, ≥95%, FG
Sigma-Aldrich
Thiolactic acid, 95%