Merck
  • Correlation between the stability and redox potential of three homologous cytochromes c from two thermophiles and one mesophile.
Correlation between the stability and redox potential of three homologous cytochromes c from two thermophiles and one mesophile.
Bioscience, biotechnology, and biochemistry (2009-02-10)
Taku Takeda, Takafumi Sonoyama, Shin-ichi J Takayama, Hajime Mita, Yasuhiko Yamamoto, Yoshihiro Sambongi
ABSTRACT
The stability of the oxidized and reduced forms of three homologous cytochromes c from two thermophiles and one mesophile was systematically monitored by means of Soret absorption measurements in the presence of various concentrations of a denaturant, guanidine thiocyanate, at pH 7.0 at 25 degrees C. Thermophilic Hydrogenobacter thermophilus cytochrome c(552) was the most stable in both redox states, followed by moderately thermophilic Hydrogenophilus thermoluteolus cytochrome c(552), and then mesophilic Pseudomonas aeruginosa cytochrome c(551). Further stability and electrochemical analysis of the three proteins and the reciprocal variants, which exhibited a different hydrophobic interaction with the heme, showed that the one with the higher stability in both redox states had the lower redox potential. Consequently, these cytochromes c probably adapted to the cellular environments of the original bacteria with correlated stability and redox potential constraints, which are in part regulated by the hydrophobicity around the heme.
Sigma-Aldrich
Guanidine thiocyanate solution, BioUltra, Molecular Biology, ~6 M in H2O
Sigma-Aldrich
Guanidine thiocyanate, BioReagent, Molecular Biology, ≥99%
Sigma-Aldrich
Guanidine thiocyanate, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
Guanidine thiocyanate, ≥97% (titration)
Sigma-Aldrich
Guanidine thiocyanate, Molecular Biology, ≥99%