Merck
Halitosis and weight loss in a cat.
Lab animal (2010-05-21)
Genaro A Coria-Avila, José L González-Hernández, Juan B Rosales-Raya, María Luisa Aguirre-Manzo, Tamara Cibrian-Llanderal, Deissy Herrera-Covarrubias, Luz Teresa Espin-Iturbe, Jorge Manzo
Product Description
Spiramycin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Spiramycin
Supelco
Spiramycin from Streptomyces sp., VETRANAL®, analytical standard, mixture of isomers