Merck
  • Experimental and spectroscopic studies of charge transfer reaction between sulfasalazine antibiotic drug with different types of acceptors.
Experimental and spectroscopic studies of charge transfer reaction between sulfasalazine antibiotic drug with different types of acceptors.
Drug testing and analysis (2011-02-16)
Moamen S Refat, Sabry A El-Korashy, Ibrahim M El-Deen, Shaima M El-Sayed
ABSTRACT
The charge-transfer (CT) interactions between the electron donor sulfasalazine (SS) and the acceptors 2,3-dichloro-5,6-dicyano-1,4-benzoquinone (DDQ), p-chloranil (CHL), picric acid (PA) and iodine have been studied spectrophotometrically in CHCl(3) or CH(3) OH solutions. The formed solid CT complexes were also isolated and characterized through infrared, (1) H-NMR, mass spectra as well as elemental and thermal analysis. The CT complexes were discussed in terms of formation constant (K(CT) ), molar extinction coefficient (ε(CT) ), standard free energy (ΔG°), oscillator strength (f), transition dipole moment (µ), resonance energy (R(N) ) and ionization potential (I(D) ). The stoichiometry of these complexes was found to be 1:1 molar ratio and having the formulae [(SS)(DDQ)], [(SS)(CHL)], [(SS)(PA)] and [(SS)(2) I](+) · I(3) (-) , respectively. The charge transfer interaction was successfully applied to the determination of SS drug using mentioned σ and π-acceptors also, the results obtained herein are satisfactory for estimation of SS compound in the pharmaceutical form.
Sigma-Aldrich
Tetrachloro-1,4-benzoquinone, 99%
Supelco
Chloranil, PESTANAL®, analytical standard
Sigma-Aldrich
2,3-Dichloro-5,6-dicyano-p-benzoquinone, 98%