Merck
  • Production of aromatic green gasoline additives via catalytic pyrolysis of acidulated peanut oil soap stock.
Production of aromatic green gasoline additives via catalytic pyrolysis of acidulated peanut oil soap stock.
Bioresource technology (2011-07-12)
R Hilten, R Speir, J Kastner, K C Das
ABSTRACT
Catalytic pyrolysis was used to generate gasoline-compatible fuel from peanut oil soap stock (PSS), a high free fatty acid feedstock, using a fixed-bed reactor at temperatures between 450 and 550°C with a zeolite catalyst (HZSM-5). PSS fed at 81 gh(-1) along with 100 mL min(-1) inert gas was passed across a 15 g catalyst bed (WHSV=5.4h(-1), gas phase residence time=34s). Results indicate that fuel properties of PSS including viscosity, heating value, and O:C ratio were improved significantly. For PSS processed at 500°C, viscosity was reduced from 59.6 to 0.9 mm(2)s(-1), heating value was increased from 35.8 to 39.3 MJL(-1), and the O:C ratio was reduced from 0.07 to 0.02. Aromatic gasoline components (e.g., BTEX), were formed in concentrations as high as 94% (v/v) in catalytically-cracked PSS with yields ranging from 22% to 35% (v/v of PSS feed).