Merck
  • Shedding light by cancer redox-human NAD(P)H:quinone oxidoreductase 1 activation of a cloaked fluorescent dye.
Shedding light by cancer redox-human NAD(P)H:quinone oxidoreductase 1 activation of a cloaked fluorescent dye.
Chemical communications (Cambridge, England) (2011-09-20)
William C Silvers, Alex S Payne, Robin L McCarley
ABSTRACT
A new quinone propionic acid-cloaked rhodamine fluorophore has its fluorescence revealed (de-cloaked) upon activation by human NAD(P)H:quinone oxidoreductase 1 (hNQO1), an upregulated enzyme in cancer cells and tumors.
Sigma-Aldrich
Morpholine, ACS reagent, ≥99.0%
Sigma-Aldrich
Morpholine, ReagentPlus®, ≥99%
Sigma-Aldrich
Morpholine, purified by redistillation, ≥99.5%
Supelco
Morpholine, analytical standard