Merck
  • International Conference on Harmonisation; final recommendation for the revision of the permitted daily exposure for the solvent cumene according to the maintenance procedures for the guidance Q3C Impurities: Residual Solvents; availability. Notice.
International Conference on Harmonisation; final recommendation for the revision of the permitted daily exposure for the solvent cumene according to the maintenance procedures for the guidance Q3C Impurities: Residual Solvents; availability. Notice.
Federal register (2012-03-03)
ABSTRACT
The Food and Drug Administration (FDA) is announcing the availability of a final recommendation for the revision of the permitted daily exposure (PDE) for the solvent cumene according to the maintenance procedures for the guidance for industry entitled ``Q3C Impurities: Residual Solvents.'' The recommendation was prepared under the auspices of the International Conference on Harmonisation of Technical Requirements for Registration of Pharmaceuticals for Human Use (ICH).
Sigma-Aldrich
Cumene, 98%
Supelco
Cumene, analytical standard
Supelco
Cumene, Pharmaceutical Secondary Standard; Certified Reference Material