Merck
  • Decolourisation of dyes under electro-Fenton process using Fe alginate gel beads.
Decolourisation of dyes under electro-Fenton process using Fe alginate gel beads.
Journal of hazardous materials (2012-03-03)
E Rosales, O Iglesias, M Pazos, M A Sanromán
ABSTRACT
This study focuses on the application of electro-Fenton technique by use of catalytic activity of Fe alginate gel beads for the remediation of wastewater contaminated with synthetic dyes. The Fe alginate gel beads were evaluated for decolourisation of two typical dyes, Lissamine Green B and Azure B under electro-Fenton process. After characterization of Fe alginate gel beads, the pH effect on the process with Fe alginate beads and a comparative study of the electro-Fenton process with free Fe and Fe alginate bead was done. The results showed that the use of Fe alginate beads increases the efficiency of the process; moreover the developed particles show a physical integrity in a wide range of pH (2-8). Around 98-100% of dye decolourisation was obtained for both dyes by electro-Fenton process in successive batches. Therefore, the process was performed with Fe alginate beads in a bubble continuous reactor. High color removal (87-98%) was attained for both dyes operating at a residence time of 30 min, without operational problems and maintaining particle shapes throughout the oxidation process. Consequently, the stable performance of Fe alginate beads opens promising perspectives for fast and economical treatment of wastewater polluted by dyes or similar organic contaminants.
Sigma-Aldrich
Lissamine Green B, Dye content 60 %
Supelco
Green S, analytical standard
Sigma-Aldrich
Azure B, prepared by direct synthesis
Sigma-Aldrich
Azure B, certified by the BSC