Merck
  • A gas chromatography-tandem quadrupole mass spectrometric analysis of policosanols in commercial vegetable oils.
A gas chromatography-tandem quadrupole mass spectrometric analysis of policosanols in commercial vegetable oils.
Journal of food science (2012-03-16)
Dong Min Jung, Mi Jin Lee, Suk Hoo Yoon, Mun Yhung Jung
ABSTRACT
Reportedly policosanols (PCs) have various beneficial functionalities on health. A gas chromatography-tandem mass spectrometry (GC-MS/MS) with a low limit of detection (LOD), and high specificity, recovery, and precision was successfully established for the PC analysis in vegetable oils. The LODs for the PCs were in the range of 0.002 to 0.016 μg/mL. The relative standard deviation (RSD) for the repeated analysis of PCs was less than 3.356%. The mean recoveries for spiked heptacosanol and octacosanol in vegetable oil were 102.3% and 106.3%, respectively. The total PC contents in the vegetable oils varied from 3.01 to 427.83 mg/kg oil. Perilla seed, grape seed, and rice bran oils were found to be highly rich sources of PCs, containing 427.83, 245.15, and 171.17 mg PCs/kg oil, respectively. Corn, sesame, and soybean oils contained only a negligible quantity of PCs. The PC composition in vegetable oils was greatly source dependent. In perilla seed oil, octacosanol was the single most predominant component, representing 55.93% of the total PC. In grape seed oil, however, hexacosanol is the most abundant PC, followed by octacosanol, tetracosanol, and triacontanol in a decreasing order. The major PCs in rice bran oil were triacontanol, octacosanol, hexacosanol, and tetracosanol, which constituted over 87.3% of the total PC. This represents the 1st report on the composition and contents of PC in most vegetable oils analyzed here. The information might be used for the development of vegetable oil products with beneficial functionality.
Sigma-Aldrich
1-Octacosanol, ≥99% (GC)
Sigma-Aldrich
1-Triacontanol, ≥98% (capillary GC)