Merck
  • Substituted phenoxypropyl-(R)-2-methylpyrrolidine aminomethyl ketones as histamine-3 receptor inverse agonists.
Substituted phenoxypropyl-(R)-2-methylpyrrolidine aminomethyl ketones as histamine-3 receptor inverse agonists.
Bioorganic & medicinal chemistry letters (2012-03-28)
Allison L Zulli, Lisa D Aimone, Joanne R Mathiasen, John A Gruner, Rita Raddatz, Edward R Bacon, Robert L Hudkins
ABSTRACT
Optimization of a series of aminomethyl ketone diamine H(3)R antagonists to reduce the brain exposure by lowering the pKa, led to molecules with improved pharmacokinetic properties. Compounds 9, 19, and 25 had high affinity for human H(3)R and demonstrated in vivo H(3)R functional activity in the rat dipsogenia model. Compound 9 displayed modest wake-promoting activity in the rat EEG/EMG model.
Sigma-Aldrich
1-Propanol, anhydrous, 99.7%
Sigma-Aldrich
1-Propanol, suitable for HPLC, ≥99.9%
Supelco
1-Propanol, analytical standard
Sigma-Aldrich
1-Propanol, ≥99%, FG
Sigma-Aldrich
1-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
1-Propanol, natural, ≥98%, FG
Sigma-Aldrich
1-Propanol, purum, ≥99.0% (GC)
Sigma-Aldrich
1-Propanol, ≥99% (GC), purum