Merck
  • Comparison of the effects of IV administration of meloxicam, carprofen, and flunixin meglumine on prostaglandin E(2) concentration in aqueous humor of dogs with aqueocentesis-induced anterior uveitis.
Comparison of the effects of IV administration of meloxicam, carprofen, and flunixin meglumine on prostaglandin E(2) concentration in aqueous humor of dogs with aqueocentesis-induced anterior uveitis.
American journal of veterinary research (2012-04-27)
Margi A Gilmour, Mark E Payton
ABSTRACT
To compare the effects of meloxicam, carprofen, and flunixin meglumine administered IV on the concentration of prostaglandin E(2) (PGE(2)) in the aqueous humor of dogs with aqueocentesis-induced anterior uveitis. 15 adult dogs with ophthalmically normal eyes. Each dog was assigned to 1 of 4 treatment groups. Treatment groups were saline (0.9% NaCl) solution (1 mL, IV), meloxicam (0.2 mg/kg, IV), carprofen (4.4 mg/kg, IV), and flunixin meglumine (0.5 mg/kg, IV). Each dog was anesthetized, treatment was administered, and aqueocentesis was performed on each eye at 30 and 60 minutes after treatment. Aqueous humor samples were frozen at -80°C until assayed for PGE(2) concentration with an enzyme immunoassay kit. For all 4 treatment groups, PGE(2) concentration was significantly higher in samples obtained 60 minutes after treatment, compared with that in samples obtained 30 minutes after treatment, which indicated aqueocentesis-induced PGE(2) synthesis. For aqueous humor samples obtained 60 minutes after treatment, PGE(2) concentration did not differ significantly among groups treated with saline solution, meloxicam, and carprofen; however, the PGE(2) concentration for the group treated with flunixin meglumine was significantly lower than that for each of the other 3 treatment groups. Flunixin meglumine was more effective than meloxicam or carprofen for minimizing the PGE(2) concentration in the aqueous humor of dogs with experimentally induced uveitis. Flunixin meglumine may be an appropriate pre-medication for use prior to intraocular surgery in dogs.
Product Description
Flunixin meglumine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Clonixin, ≥98% (HPLC)
Sigma-Aldrich
Flunixin meglumine, ≥98% (HPLC)
Supelco
Carprofen, VETRANAL®, analytical standard