Merck
Rh2O3 versus IrO2: relativistic effects and the stability of Ir4+.
Journal of physics. Condensed matter : an Institute of Physics journal (2012-05-01)
M-S Miao, R Seshadri
ABSTRACT
Despite the wide-ranging applications of binary Rh and Ir oxides, their stability and trends in Rh and Ir oxidation states are not fully understood. Using first-principles electronic structure calculations, we demonstrate that the origin of the categorical stability of Ir(4+) is the relativistic contraction of the 6s orbital and, consequently, an expansion of 5d orbitals. Relativistic effects significantly stabilize Ir(4+)-containing metallic rutile IrO(2) over a wide range of O chemical potentials, despite the choice that Ir has of forming semiconducting corundum Ir(2)O(3). In contrast, Rh is found to display a wider stability range for corundum Rh(2)O(3) with Rh(3+) and a greater propensity for multiple oxidation states.
Sigma-Aldrich
Sodium perrhenate, 99.99% trace metals basis
Sigma-Aldrich
Ammonium perrhenate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium perrhenate, ≥99%
Sigma-Aldrich
Potassium perrhenate, 99.98% trace metals basis
Sigma-Aldrich
Iridium(IV) oxide, 99.9% trace metals basis
Sigma-Aldrich
Potassium perrhenate, 99%