Merck
  • Determination of lapachol in the presence of other naphthoquinones using 3MPA-CdTe quantum dots fluorescent probe.
Determination of lapachol in the presence of other naphthoquinones using 3MPA-CdTe quantum dots fluorescent probe.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-05-18)
Ricardo Q Aucélio, Ana I Peréz-Cordovés, Juliano L Xavier Lima, Aurélio Baird B Ferreira, Ana M Esteva Guas, Andrea R da Silva
ABSTRACT
3MPA-CdTe QDs in aqueous dispersion was used as a fluorescent probe for the determination of lapachol, a natural naphthoquinone found in plants of the Bignoniaceae family genus Tabebuia. Working QDs dispersions (4.5×10(-8) mol L(-1) of QDs) was prepared in aqueous media containing Tris-HCl buffer pH 7.4 and methanol (10% in volume). The excitation was made at 380 nm with signal measurement at 540 nm. To establish a relationship between fluorescence (corrected to inner filter effect) and concentration of lapachol, a Stern-Volmer model was used. The linear range obtained was from 1.0×10(-5) to 1.0×10(-4) mol L(-1). The limit of detection (x(b)-3s(b)) was 8.0×10(-6) mol L(-1). The 3MPA-CdTe QDs probe was tested in the determination of lapachol in urine, previously cleansed in an acrylic polymer. The average recovery was satisfactory.
Sigma-Aldrich
Lapachol, 98%
Sigma-Aldrich
3-Mercaptopropionic acid, ≥99.0% (HPLC)
Sigma-Aldrich
3-Mercaptopropionic acid, ≥99%