Merck
[Antioxidant properties of proteins after freezing-thawing].
Ukrains'kyi biokhimichnyi zhurnal (1999 ) (2012-06-12)
S L Rozanova, E D Rozanova, O A Nardid
ABSTRACT
Experimental data are presented which were obtained under comparative evaluation of influence of different freezing-thawing conditions on antioxidant properties of isolated proteins: human serum albumin, cytochrome c from the horse heart and glucose oxidase from Aspergillus niger. The observed protein antioxidant activity alterations are assumed to be a result of protein conformational changes. The character of freezing-thawing influence on the protein antioxidant activity depends on the molecular structure and cooling conditions.
Sigma-Aldrich
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p′-disulfonic acid monosodium salt hydrate, 97%
Sigma-Aldrich
Iron(II) chloride, 98%
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Iron(II) chloride tetrahydrate, 99.99% trace metals basis
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Supelco
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4′,4′′-disulfonic acid sodium salt, for spectrophotometric det. of Fe, 97.0%
Sigma-Aldrich
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4′,4′′-disulfonic acid sodium salt, BioXtra
Sigma-Aldrich
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4′,4′′-disulfonic acid sodium salt, Indicator ligand
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Iron(II) chloride tetrahydrate, ReagentPlus®, 98%
Sigma-Aldrich
Iron(II) chloride tetrahydrate, puriss. p.a., ≥99.0% (RT)