Merck
[Study on the bioactive constituents of Piper wallichii].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2012-06-28)
Guo-Wei Zhao, Wen Xia, Ping Chen, En-Ji Han, Lan Xiang
ABSTRACT
To investigate the bioactive constituents in the stem of Piper wallichii. Compounds were separated by column chromatography of silica gel, ODS-A and Sephadex LH-20. Their structures were elucidated based on spectral analysis. DPPH scavenging activity and AchE inhibitory activity were tested. 10 compounds were isolated and their structures were identified as 3,4-methylenedioxy-benzoic acid (1), vanillic acid (2), benzoic acid (3), N-p-coumaroyltyramine (4), futoenone (5), futoquinol (6), isofutoquinol A (7), 4-hydroxy-3,5-dimethoxy-benzoic acid (8), futoamide (9), dihydropiperlonguminine (10). Compounds 1-6 are isolated from P. wallichii for the first time. Vanillic acid (2) and 4-hydroxy-3,5-dimethoxy-benzoic acid (8) show scavenging activity against DPPH radical with ED50 at 224.33 microg/mL and 11.44 microg/mL, respectively. No compound shows inhibition activity against AchE.
Sigma-Aldrich
Vanillic acid, ≥97%, FG
Sigma-Aldrich
Vanillic acid, purum, ≥97.0% (HPLC)
Sigma-Aldrich
Vanillic acid, 97%
Supelco
Vanillic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Piperonylic acid, 99%