Merck
  • [Influence of different solutions on extracting of active components in Danggui Chishao drug pair].
[Influence of different solutions on extracting of active components in Danggui Chishao drug pair].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2012-07-17)
Wen Ding, Dawei Qian, Pei Liu, Jin'ao Duan, Erxin Shang, Yuping Tang, Jing Wu
ABSTRACT
To study the dissolution rate of active components of different extracting solvents of Danggui Chishao drug pair. The dissolution rates of phenolic acids (ferulic acid, vanillic acid and gallic acid), monoterpenes (gallic acid, peoniflorin, albiflorin, hydroxypeoniflorin and galloylpaeoniflorin) and phthalates (senkyunolide and ligustilide) contained in Danggui Chishao drug pair were determined by quadrupole time-of-flight tandem mass spectrometry (UPLC-Q-TOF-MS) and ultra high performance liquid chromatography in combination with triple-quadrupole mass spectrometry (UPLC-TQ-MS). The dissolution rates of phenolic acids and monoterpenes decreased with the increase in alcohol concentration, on the contrary the rates of phthalates increased. The relative dissolution rates of most active components were relatively high in water and low concentration alcohol than other solvents. Liquid chromatography-mass spectrometry is practical for comprehensive multi-component assessment on traditional Chinese medicine preparation processes and can provide reference for optimization of processing parameters.
Sigma-Aldrich
Vanillic acid, ≥97%, FG
Sigma-Aldrich
Vanillic acid, purum, ≥97.0% (HPLC)
Sigma-Aldrich
Vanillic acid, 97%
Supelco
Vanillic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Albiflorin, ≥95% (LC/MS-ELSD)
Sigma-Aldrich
Paeoniflorin, ≥98% (HPLC)