Merck
  • Supramolecular architecture based on the self-assembling of multiwall carbon nanotubes dispersed in polyhistidine and glucose oxidase: Characterization and analytical applications for glucose biosensing.
Supramolecular architecture based on the self-assembling of multiwall carbon nanotubes dispersed in polyhistidine and glucose oxidase: Characterization and analytical applications for glucose biosensing.
Biosensors & bioelectronics (2012-07-17)
Pablo R Dalmasso, María L Pedano, Gustavo A Rivas
ABSTRACT
We report for the first time the development of a sensitive and selective glucose biosensor based on the self-assembling of multiwall carbon nanotubes (MWCNTs) dispersed in polyhistidine (Polyhis) and glucose oxidase (GOx) on glassy carbon electrodes (GCE). The supramolecular architecture was characterized by SEM, FT-IR and electrochemical techniques. The optimum multistructure was obtained with five (MWCNT-Polyhis/GOx) bilayers and one layer of Nafion as anti-interferent barrier. The sensitivity at 0.700V was (1.94±0.03) mAM(-1) (r=0.9991), with a linear range between 0.25 and 5.00mM, a detection limit of 2.2μM and a quantification limit of 6.7μM with minimum interference from lactose (1.5%), maltose (5.7%), galactose (1.2%), ascorbic acid (1.0%), and uric acid (3.3%). The biocatalytic layer demonstrated to be highly reproducible since the R.S.D. for 10 successive amperometric calibrations using the same surface was 3.6%. The sensitivity of the biosensor after 15 day storage at 4°C remained at 90% of its original value. The combination of the excellent dispersing properties and polycationic nature of polyhistidine, the stability of the MWCNT-Polyhis dispersion, the electrocatalytic properties of MWCNTs, the biocatalytic specificity of GOx, and the permselective properties of Nafion have allowed building up a sensitive, selective, robust, reproducible and stable glucose amperometric biosensor for the quantification of glucose in milk samples.
Sigma-Aldrich
Poly-L-histidine, mol wt 5,000-25,000
Pricing and availability is not currently available.