Merck
  • Evaluation of Clearview Exact PBP2a, a new immunochromatographic assay, for detection of low-level methicillin-resistant Staphylococcus aureus (LL-MRSA).
Evaluation of Clearview Exact PBP2a, a new immunochromatographic assay, for detection of low-level methicillin-resistant Staphylococcus aureus (LL-MRSA).
Journal of clinical microbiology (2012-07-21)
Claire Nonhoff, Sandrine Roisin, Marie Hallin, Olivier Denis
ABSTRACT
We evaluated the performance of a new immunochromatographic assay (ICA), the Clearview Exact PBP2a, for rapid detection of penicillin-binding protein 2a (PBP2a) in a challenge set of Staphylococcus aureus strains showing MICs to oxacillin of ≤16 mg/liter. The sensitivity and specificity of the ICA were 96.6% and 100%, respectively.
Product Description
Supelco
Oxacillin sodium salt monohydrate, VETRANAL®, analytical standard
Sigma-Aldrich
Oxacillin sodium salt, 815-950 μg/mg (Oxacillin)