Merck
  • Cutaneous adverse reactions to amoxicillin-clavulanic acid suspension in children: the role of sodium benzoate.
Cutaneous adverse reactions to amoxicillin-clavulanic acid suspension in children: the role of sodium benzoate.
Current drug safety (2012-08-10)
Francesca Mori, Simona Barni, Neri Pucci, Maria Elisabetta Rossi, Maurizio de Martino, Elio Novembre
ABSTRACT
In Europe amoxicillin plus clavulanic acid is the most commonly prescribed antibiotic and sodium benzoate is contained in the suspension formulation as a preservative. We studied the relevance of sodium benzoate as the culprit agent. In a group of children with a history of adverse reactions to amoxicillin plus clavulanic acid suspension. A total of 89 children were enrolled over a period of 3 years (2006 - 2009). Single blind oral provocation tests (OPTs) with amoxicillin plus clavulanic acid, sodium benzoate and placebo were performed. 20 children with recurrent idiopathic urticaria were investigated as a control group. according to personal history: 70% of reactions were late in developing while 23% of reactions were immediate and for 5% of the cases it was not possible to define the timing. 8 children (8/89=9%) resulted positive to the provocation tests with amoxicillin plus clavulanic acid; ten children (10/89=11%) had positive results with sodium benzoate; 3% had a double positivity (i.e. excipient and active drug). The timing of reactions significantly differs between the Amoxicillin plus clavulanic acid and sodium benzoate groups (p=0.002). Sodium benzoate probably acts through a non-immunologic mechanism and care should be given to children allergic to sodium benzoate containing pharmaceutical formulations.
Product Description
Supelco
Sodium benzoate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium benzoate, BioXtra, ≥99.5%
Sigma-Aldrich
Sodium benzoate, puriss., meets analytical specification of Ph. Eur., BP, FCC, E211, 99.0-100.5% (calc. to the dried substance), powder
Sigma-Aldrich
Sodium benzoate, purum p.a., ≥99.0% (NT)
Sigma-Aldrich
Sodium benzoate, ReagentPlus®, 99%