Merck
  • 2-Amino-1,3,4-thiadiazole derivative (FABT) inhibits the extracellular signal-regulated kinase pathway and induces cell cycle arrest in human non-small lung carcinoma cells.
2-Amino-1,3,4-thiadiazole derivative (FABT) inhibits the extracellular signal-regulated kinase pathway and induces cell cycle arrest in human non-small lung carcinoma cells.
Bioorganic & medicinal chemistry letters (2012-08-11)
M Juszczak, J Matysiak, M Szeliga, P Pożarowski, A Niewiadomy, J Albrecht, W Rzeski
ABSTRACT
The anticancer potential of 2-amino-1,3,4-thiadiazole compounds has been documented by in vitro and in vivo studies. In our previous research, we described the synthesis as well as the antiproliferative and neuroprotective activities of 2-(4-fluorophenyloamino)-5-(2,4-dihydroxyphenyl)-1,3,4-thiadiazole (FABT). The aim of the present study was to investigate the molecular mechanisms involved in FABT-induced growth inhibition in A549 lung carcinoma cells. Western blotting analysis revealed that FABT inhibited the activation of the extracellular signal-regulated kinase 1/2 (ERK1/2) pathway, and Real-time PCR analysis showed no changes in the expression of P44ERK1 and CREB1 genes. Furthermore, FABT induced cell cycle arrest in the GO/G1 phase and enhanced p27/Kip1 expression. Our results suggest that FABT acts by inhibiting ERK1/2 pathway and cell cycle progression through G1 into S phase in A549 cells. Further studies are needed to completely explain the molecular mechanisms of anticancer action of this 2-aminothiadiazole derivative.
Sigma-Aldrich
2-Amino-1,3,4-thiadiazole, 97%