Merck
  • A single measurement of serum phylloquinone is an adequate indicator of long-term phylloquinone exposure in healthy older adults.
A single measurement of serum phylloquinone is an adequate indicator of long-term phylloquinone exposure in healthy older adults.
The Journal of nutrition (2012-08-24)
Nancy Presse, Pierrette Gaudreau, Carol E Greenwood, Marie-Jeanne Kergoat, Jose A Morais, Hélène Payette, Bryna Shatenstein, Guylaine Ferland
ABSTRACT
Assessment of long-term phylloquinone exposure is challenging in studies investigating vitamin K in health. Data are equivocal as to whether a single measurement of circulating phylloquinone would be adequate. The primary purpose of the present study was to validate the use of a single measurement of serum phylloquinone as a surrogate for long-term phylloquinone exposure in healthy older adults. Using data from the Québec Longitudinal Study on Nutrition and Successful Aging, the objectives were to: 1) determine the reproducibility of circulating phylloquinone over 2 y (n = 234); 2) calculate how a single measurement would rank or classify individuals and attenuate the regression coefficient between circulating phylloquinone and a health outcome; and 3) investigate the association of a single measurement of serum phylloquinone with long-term phylloquinone intakes assessed over the year prior to the blood draw (n = 228). The variance analysis based on 2 blood samples showed a fair to good reproducibility for serum phylloquinone (intra-class correlation = 0.49). The correlation coefficient between the ranking of individuals based on a single measurement of circulating phylloquinone and the "true" ranking would be 0.70. The multiple regression analysis showed that long-term phylloquinone intake was the strongest predictor of serum phylloquinone (t = 4.94; P < 0.001). The partial correlation coefficient (r = 0.32) was comparable with those reported in studies where blood sampling and diet recording were juxtaposed and/or multiple blood samples were used. The present study provides evidence that the use of a single measurement of circulating phylloquinone is adequate for assessing long-term phylloquinone exposure in healthy older adults.
Product Description
Supelco
Phylloquinone (K1), analytical standard
Supelco
Phytonadione, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Vitamin K1, BioXtra, ≥99.0% (sum of isomers, HPLC), mixture of isomers
Sigma-Aldrich
Vitamin K1, viscous liquid
Phytomenadione, European Pharmacopoeia (EP) Reference Standard