Merck
  • Nitrous-acid-mediated synthesis of iron-nitrosyl-porphyrin: pH-dependent release of nitric oxide.
Nitrous-acid-mediated synthesis of iron-nitrosyl-porphyrin: pH-dependent release of nitric oxide.
Chemistry, an Asian journal (2012-08-29)
Jagannath Bhuyan, Sabyasachi Sarkar
ABSTRACT
Two iron-nitrosyl-porphyrins, nitrosyl[meso-tetrakis(3,4,5-trimethoxyphenylporphyrin]iron(II) acetic acid solvate (3) and nitrosyl[meso-tetrakis(4-methoxyphenylporphyrin]iron(II) CH(2)Cl(2) solvate (4), were synthesized in quantitative yield by using a modified procedure with nitrous acid, followed by oxygen-atom abstraction by triphenylphosphine under an argon atmosphere. These nitrosyl porphyrins are in the {FeNO}(7) class. Under an argon atmosphere, these compounds are relatively stable over a broad range of pH values (4-8) but, under aerobic conditions, they release nitric oxide faster at high pH values than that at low pH values. The generated nitric-oxide-free iron(III)-porphyrin can be re-nitrosylated by using nitrous acid and triphenylphosphine. The rapid release of NO from these Fe(II) complexes at high pH values seems to be similar to that in nitrophorin, a nitric-oxide-transport protein, which formally possesses Fe(III). However, because the release of NO occurs from ferrous-nitrosyl-porphyrin under aerobic conditions, these compounds are more closely related to nitrobindin, a recently discovered heme protein.
Sigma-Aldrich
Triphenylphosphine, ≥95.0% (GC)
Sigma-Aldrich
Triphenylphosphine, ReagentPlus®, 99%