Merck
  • Tauroursodeoxycholic acid affects PPARγ and TLR4 in Steatotic liver transplantation.
Tauroursodeoxycholic acid affects PPARγ and TLR4 in Steatotic liver transplantation.
American journal of transplantation : official journal of the American Society of Transplantation and the American Society of Transplant Surgeons (2012-09-22)
M B Jiménez-Castro, M Elias-Miro, M Mendes-Braz, A Lemoine, A Rimola, J Rodés, A Casillas-Ramírez, C Peralta
ABSTRACT
Numerous steatotic livers are discarded for transplantation because of their poor tolerance to ischemia-reperfusion (I/R). We examined whether tauroursodeoxycholic acid (TUDCA), a known inhibitor of endoplasmic reticulum (ER) stress, protects steatotic and nonsteatotic liver grafts preserved during 6 h in University of Wisconsin (UW) solution and transplanted. The protective mechanisms of TUDCA were also examined. Neither unfolded protein response (UPR) induction nor ER stress was evidenced in steatotic and nonsteatotic liver grafts after 6 h in UW preservation solution. TUDCA only protected steatotic livers grafts and did so through a mechanism independent of ER stress. It reduced proliferator-activated receptor-γ (PPARγ) and damage. When PPARγ was activated, TUDCA did not reduce damage. TUDCA, which inhibited PPARγ, and the PPARγ antagonist treatment up-regulated toll-like receptor 4 (TLR4), specifically the TIR domain-containing adaptor inducing IFNβ (TRIF) pathway. TLR4 agonist treatment reduced damage in steatotic liver grafts. When TLR4 action was inhibited, PPARγ antagonists did not protect steatotic liver grafts. In conclusion, TUDCA reduced PPARγ and this in turn up-regulated the TLR4 pathway, thus protecting steatotic liver grafts. TLR4 activating-based strategies could reduce the inherent risk of steatotic liver failure after transplantation.
Sigma-Aldrich
Sodium taurochenodeoxycholate
Pricing and availability is not currently available.