Merck
  • The effect of ligustrazine on L-type calcium current, calcium transient and contractility in rabbit ventricular myocytes.
The effect of ligustrazine on L-type calcium current, calcium transient and contractility in rabbit ventricular myocytes.
Journal of ethnopharmacology (2012-10-13)
Zhiqiang Ren, Jihua Ma, Peihua Zhang, Antao Luo, Shuo Zhang, Linghao Kong, Chunping Qian
ABSTRACT
Ligustrazine, the biologically active ingredient isolated from a popular Chinese medicinal plant, Ligusticum chuanxiong Hort. (Umbelliferae), has been used effectively to treat ischemic heart diseases, cerebrovascular and thrombotic vascular diseases since the 1970s. At present, the effect of ligustrazine on L-type calcium current (I(Ca-L)) of ventricular myocytes remains controversial. In this study, we use the whole-cell patch-clamp techniques and video-based edge detection and dual excitation fluorescence photomultiplier systems to study the effects of ligustrazine on I(Ca-L), and calcium transient and contractility in rabbit ventricular myocytes in the absence and presence of isoprenaline (ISO). Ligustrazine (5 μM) in low concentration did not affect I(Ca-L) (P>0.05), higher concentrations of this drug (10, 20, 40, 80 μM) inhibited I(Ca-L) in a concentration-dependent manner and reduced I(Ca-L) by 9.6 ± 2.9%, 21.0 ± 4.3%, 33.9 ± 4.3%, and 51.6 ± 7.3%, respectively. Under normal conditions, ligustrazine (40 μΜ) reduced baseline of fura-2 fluorescence intensities (FFI, 340/380 ratio), namely diastolic calcium concentration, changes in FFI (ΔFFI, 340/380 ratio) and maximal velocity of Ca(2+) rise and decay (340/380 ratio/ms) by 6.3%, 26.1%, 25.2%, and 26.5%, and decreased sarcomere peak shorting (PS) and maximal velocity of shorting and relengthening by 36.4%, 31.9%, and 25.0%, respectively. Similarly, ligustrazine (40 μM) reduced baseline FFI, ΔFFI, and maximal velocity of Ca(2+) rise and decay by 14.1%, 51.1%, 35.2%, and 41.1%, and reduced sarcomere PS and maximal velocity of shorting and relengthening by 38.6%, 50.0% and 39.1%, respectively, in the presence of ISO. Ligustrazine not only significantly inhibits I(Ca-L) in a concentration-dependent manner but also suppressed calcium transient and contraction in the absence and presence of ISO.
Sigma-Aldrich
2,3,5,6-Tetramethylpyrazine, natural, ≥98%, FG
Sigma-Aldrich
2,3,5,6-Tetramethylpyrazine, 98%
Sigma-Aldrich
2,3,5,6-Tetramethylpyrazine, ≥98%, FG
Supelco
2,3,5,6-Tetramethylpyrazine, analytical standard