Merck
  • Evaluation of anti-nociceptive, anti-inflammatory and antipyretic activities of Artemisia scoparia hydromethanolic extract.
Evaluation of anti-nociceptive, anti-inflammatory and antipyretic activities of Artemisia scoparia hydromethanolic extract.
Journal of ethnopharmacology (2012-11-03)
Muheet Habib, Ishrat Waheed
ABSTRACT
Artemisia scoparia (redstem wormwood) locally known as jhahoo or jaukay, is traditionally used in pain, inflammation and febrile conditions. So far, little or no scientific work has been reported to validate its folk uses in the alleviation of pain, fever and inflammation. The present study was designed to explore the analgesic, anti-inflammatory and antipyretic effects of the Artemisia scoparia hydromethanolic extract (ASHME), and to validate its traditional use in Asia. This study made use of thermal (hot plate induced) and chemical (acetic acid induced) nociception models in mice. In addition, the mechanism of antinociception in hot plate test was further evaluated in the presence of caffeine (10mg/kg), naloxone (2mg/kg) and monosodium glutamate (1g/kg). While carrageenan induced rat paw edema and yeast induced mouse pyrexia models were used to test the anti-inflammatory and antipyretic activities. Administration of single intraperitoneal doses (400mg/kg and 800 mg/kg) of ASHME significantly reduced the carrageenan induced paw edema in rats (P<0.05, P<0.001) by 54% and 74%, increased the thermal nociception time in the hot plate test up to 2- and 2.5-fold (P<0.01, P<0.001), inhibited the acetic acid induced writhings in mice by 41.12% and 61.53% (P<0.001), and attenuated the yeast induced pyrexia in mice by nearly 74% and 90% respectively (P<0.01, P<0.001). Caffeine (10mg/kg), naloxone (2mg/kg) and monosodium glutamate (1g/kg) significantly (P<0.001) abolished the anti-nociceptive response of ASHME (400mg/kg). These findings suggest that the Artemisia scoparia hydromethanolic extract of ASHME possesses anti-nociceptive, anti-inflammatory and antipyretic potentials, which support its use, for the said conditions, in traditional medicine and should be further exploited for its use in clinical medicine.
Sigma-Aldrich
L-Glutamic acid monosodium salt hydrate, ≥99% (HPLC), powder
Sigma-Aldrich
L-Glutamic acid monosodium salt hydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%