Merck
  • Edaravone, a novel free radical scavenger, prevents steroid-induced osteonecrosis in rabbits.
Edaravone, a novel free radical scavenger, prevents steroid-induced osteonecrosis in rabbits.
Rheumatology (Oxford, England) (2012-11-23)
Guang-Yi Li, Yong Feng, Tak S Cheng, Ji-Min Yin, Chang-Qing Zhang
ABSTRACT
To investigate the efficacy of edaravone, a novel free radical scavenger, on preventing steroid-induced osteonecrosis (ON) in a rabbit model. Thirty-six New Zealand white rabbits were divided into control (C; n = 6), steroid-administered (S; n = 15) and edaravone-administered groups (E; n = 15) after receiving an established protocol of steroid-induced ON. Before and after steroid administration, plasma levels of reduced glutathione (GSH) and lipid peroxidation (LPO) were measured for oxidative stress. Two weeks later bilateral proximal femurs were dissected for micro-CT-based micro-angiography, and the presence or absence of ON and intravascular thrombi were examined histopathologically. Immunohistochemical examination of oxidative injury in bone tissue was conducted using the anti-8-hydoxy-2'-deoxyguanosine and anti-malondialdehyde mAbs. The incidence of ON in the E group (20%) was significantly lower than in the S group (73%). Three to five days after steroid administration, the plasma GSH level was significantly higher and LPO level was significantly lower in the E group than the S group. Compared with the S group, there were significantly more small-sized perfusion vessels and fewer large-sized dilated vessels in the E group. Thrombosis incidence was significantly lower in the E group than the S group. Intraosseous vessels and haematopoietic cells that sustained oxidative injury were significantly fewer in the E group than the S group. Edaravone exerted beneficial effects on reducing incidence of steroid-induced ON by suppressing the accumulation of lipid peroxidative products and oxidative DNA damage in endothelial cells and haematopoietic cells.
Product Description
Supelco
Antipyrine, analytical standard
Sigma-Aldrich
3-Methyl-1-phenyl-2-pyrazoline-5-one, 99%