Merck
  • Gold nanocluster-based electrochemically controlled fluorescence switch surface with prussian blue as the electrical signal receptor.
Gold nanocluster-based electrochemically controlled fluorescence switch surface with prussian blue as the electrical signal receptor.
Chemical communications (Cambridge, England) (2012-11-24)
Lihua Jin, Youxing Fang, Li Shang, Yaqing Liu, Jing Li, Li Wang, Peng Hu, Shaojun Dong
ABSTRACT
A highly robust electrochemically controlled fluorescence switch based on ultrasmall Au nanoclusters has been designed by the aid of the electrochemical redox reaction of prussian blue.
Sigma-Aldrich
Iron(III) ferrocyanide