Merck
  • Structure-dependent lipid peroxidation by photoirradiation of pyrene and its mono-substituted derivatives.
Structure-dependent lipid peroxidation by photoirradiation of pyrene and its mono-substituted derivatives.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering (2012-12-19)
Tracie Perkins Fullove, Britney Johnson, Hongtao Yu
ABSTRACT
Pyrene, one of the most studied polycyclic aromatic hydrocarbons, can damage biological macromolecules and cause toxicity when irradiated by light. The effect of substituents, 1-amino, 1-hydroxy, 1-nitro, and 1-bromo, on light-induced lipid peroxidation is studied. Degradation kinetics and photoproduct analyses were conducted to test how these substituents affect the photoreaction. All five compounds have widely different photodegradation rates, with degradation half-lives, ranging from 8 min to 495 min. These rates parallel their light absorptivity. Four out of the five compounds induce lipid peroxidation when irradiated with UVA light, whereas 1-aminopyrene causes minimum or no lipid peroxidation. The relative amount of lipid peroxidation caused is: 1-bromopyrene > pyrene > 1-nitropyrene ≈ 1-hydroxypyrene > 1-aminopyrene. This relative lipid peroxidation is dependent on the substituent due to the following factors: light absorptivity, relative rates of the competing processes in the excited states, nature of the photoreaction, and nature of the photoproducts.
Sigma-Aldrich
Pyrene, sublimed grade, 99%
Sigma-Aldrich
1-Bromopyrene, 96%
Sigma-Aldrich
Pyrene, puriss. p.a., suitable for fluorescence, ≥99.0% (GC)
Supelco
Pyrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Pyrene, 98%
Supelco
Pyrene, analytical standard
Pyrene, BCR®, certified reference material