Merck
  • Copper-boosting compounds: a novel concept for antimycobacterial drug discovery.
Copper-boosting compounds: a novel concept for antimycobacterial drug discovery.
Antimicrobial agents and chemotherapy (2012-12-21)
Alexander Speer, Tej B Shrestha, Stefan H Bossmann, Randall J Basaraba, Gregory J Harber, Suzanne M Michalek, Michael Niederweis, Olaf Kutsch, Frank Wolschendorf
ABSTRACT
We and others recently identified copper resistance as important for virulence of Mycobacterium tuberculosis. Here, we introduce a high-throughput screening assay for agents that induce a copper hypersensitivity phenotype in M. tuberculosis and demonstrate that such copper-boosting compounds are effective against replicating and nonreplicating M. tuberculosis strains.
Sigma-Aldrich
Neocuproine, ≥98%
Sigma-Aldrich
Bathocuproine, 96%
Sigma-Aldrich
Bathocuproine, sublimed grade, 99.99% trace metals basis