Merck
  • Synthesis and antibacterial evaluation of typharin analog: 6,8-dihydroxy-7-methyl-3-styryl-3,4-dihydroisocoumarin.
Synthesis and antibacterial evaluation of typharin analog: 6,8-dihydroxy-7-methyl-3-styryl-3,4-dihydroisocoumarin.
Journal of Asian natural products research (2013-01-18)
Aamer Saeed, Hummera Rafique, Zaman Ashraf
ABSTRACT
Synthesis of the 6-hydroxy-7-methyl analog of typharin (8-dihydroxy-3-styryl-3,4-dihydroisocoumarin) isolated from the rhizomes of Typha capensis has been described. Direct condensation of 3,5-dimethoxy-4-methylhomophthalic acid (1) with cinnamoyl chloride at elevated temperature under inert conditions afforded 6,8-dihydroxy-7-methyl-3-styrylisocoumarin (2). Hydrolysis of isocoumarin to keto acid (3) followed by reduction and cyclodehydration of hydroxy acid analog (4) afforded ( ± )-6,8-dimethoxy-3-(4-methoxyphenyl)-3,4-dihydroisocoumarin (5). Demethylation of the latter using anhydrous aluminum chloride/ethane thiol furnished the title isocoumarin (6). Compounds (1-6) were screened for in vitro antibacterial activity against a representative panel of Gram-positive and Gram-negative bacteria using levofloxacin as the reference drug.
Product Description
Supelco
Ofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ofloxacin, fluoroquinolone antibiotic
Sigma-Aldrich
Levofloxacin, 98.0-102.0% anhydrous basis (HPLC)
Supelco
Levofloxacin, analytical standard
Supelco
Ofloxacin, VETRANAL®, analytical standard
Ofloxacin, European Pharmacopoeia (EP) Reference Standard