Merck
  • Optimization of clean fractionation process applied to switchgrass to produce pulp for enzymatic hydrolysis.
Optimization of clean fractionation process applied to switchgrass to produce pulp for enzymatic hydrolysis.
Bioresource technology (2013-01-24)
Grzegorz Brudecki, Iwona Cybulska, Kurt Rosentrater
ABSTRACT
The purpose of this study was to fractionate switchgrass (SG) to obtain hemicellulose-, lignin-rich fractions and highly digestible pulp, using a clean fractionation (CF) approach. The main objective was to produce highest glucose yield in the enzymatic hydrolysis of pulp. Effects of processing factors such as time (10-50 min), temperature (120-160 °C), catalyst concentration (0.21-0.93% w/w sulfuric acid) and organic solvent mixture composition (7-43% w/w methyl isobutyl ketone) were evaluated. Response surface methodology and central composite design were used for process optimization and statistical analyses. High lignin (75-93%) and xylan (83-100%) removal from biomass were obtained, leaving solid pulp rich in glucan (78-94%). High enzymatic hydrolysis glucose yields (more than 90%) were obtained for selected optimal conditions. Pulp can be used for ethanol production while separated xylan and lignin fractions can be used as a feedstock for value-added products which suggests the applicability of clean fractionation technology in a biorefinery concept.
Product Description
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%