Merck
  • A simple but accurate potential for the naphthalene-argon complex: applications to collisional energy transfer and matrix isolated IR spectroscopy.
A simple but accurate potential for the naphthalene-argon complex: applications to collisional energy transfer and matrix isolated IR spectroscopy.
The Journal of chemical physics (2013-01-25)
F Calvo, Cyril Falvo, Pascal Parneix
ABSTRACT
An explicit polarizable potential for the naphthalene-argon complex has been derived assuming only atomic contributions, aiming at large scale simulations of naphthalene under argon environment. The potential was parametrized from dedicated quantum chemical calculations at the CCSD(T) level, and satisfactorily reproduces available structural and energetic properties. Combining this potential with a tight-binding model for naphthalene, collisional energy transfer is studied by means of dedicated molecular dynamics simulations, nuclear quantum effects being accounted for in the path-integral framework. Except at low target temperature, nuclear quantum effects do not alter the average energies transferred by the collision or the collision duration. However, the distribution of energy transferred is much broader in the quantum case due to the significant zero-point energy and the higher density of states. Using an ab initio potential for the Ar-Ar interaction, the IR absorption spectrum of naphthalene solvated by argon clusters or an entire Ar matrix is computed via classical and centroid molecular dynamics. The classical spectra exhibit variations with growing argon environment that are absent from quantum spectra. This is interpreted by the greater fluxional character experienced by the argon atoms due to vibrational delocalization.
Sigma-Aldrich
Argon, ≥99.998%
Supelco
Naphthalene solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Naphthalene, 99%
Supelco
Naphthalene, analytical standard
Sigma-Aldrich
Naphthalene, suitable for scintillation, ≥99%
Supelco
Naphthalene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Melting point standard 79-81°C, analytical standard