Merck
  • Children presenting with calcified disc herniation: a self-limiting process.
Children presenting with calcified disc herniation: a self-limiting process.
Journal of radiology case reports (2013-02-05)
Chang Ho, Stanley Chang, Daniel Fulkerson, Jodi Smith
ABSTRACT
We present two cases of disc herniation associated with juvenile intervertebral disc calcification, a rare, self-limiting process which typically resolves with conservative treatment. Recognition of this entity prevents unnecessary diagnostic workup and possible surgical intervention. A review of the literature for this rare entity is discussed.
Product Description
Supelco
Ibuprofen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
(S)-(+)-Ibuprofen, ReagentPlus®, 99%
Ibuprofen, European Pharmacopoeia (EP) Reference Standard
Supelco
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ibuprofen
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Supelco
Ibuprofen sodium salt, ≥98% (GC)
Ibuprofen for peak identification, European Pharmacopoeia (EP) Reference Standard