Merck
  • Factors affecting the separation and bioactivity of red clover (Trifolium pratense) extracts assayed against Clostridium sticklandii, a ruminal hyper ammonia-producing bacterium.
Factors affecting the separation and bioactivity of red clover (Trifolium pratense) extracts assayed against Clostridium sticklandii, a ruminal hyper ammonia-producing bacterium.
Natural product communications (2013-02-19)
Isabelle A Kagan, Michael D Flythe
ABSTRACT
Red clover (Trifolium pratense) is rich in phenolic compounds. Both the crude phenolic extract and biochanin A, an isoflavonoid component of the extract, suppress growth of Clostridium sticklandii, a bovine, Gram-positive, ruminal hyper-ammonia producing bacterium (HAB). The purpose of this study was to determine if other components of red clover extract contributed to its anti-HAB activity. Extracts of the Kenland cultivar of red clover, collected 0 h and 24 h after cutting, were separated by normal-phase thin-layer chromatography (TLC) in either ethyl acetate-hexanes (9:1, v/v) or ethyl acetate-methanol (4:1, v/v). Bands on TLC plates were assayed by either overlaying the plates with agar seeded with C. sticklandii, or setting the excised bands face-down onto plates of bacteria-seeded agar. Biochanin A inhibited C. sticklandii growth on TLC plates when as little as 8 nmol was present in the extract. An antimicrobial band, seen in a previous bioassay, was not found, suggesting that this second compound may be more labile than biochanin A.
Sigma-Aldrich
Biochanin A
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)
Supelco
Genistein, analytical standard