Merck
  • Sensitive and selective detection of trace copper in standard alloys, food and biological samples using a bulk optode based on N,N'-(4,4'-ethylene biphenyl) bis(3-methoxy salicylidine imine) as neutral carrier.
Sensitive and selective detection of trace copper in standard alloys, food and biological samples using a bulk optode based on N,N'-(4,4'-ethylene biphenyl) bis(3-methoxy salicylidine imine) as neutral carrier.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2013-02-26)
Majid Arvand, Zahra Lashkari
ABSTRACT
In this paper, an optical sensing film has been proposed for sensitive determination of copper (ІІ) ion in aqueous solutions. The copper sensing membrane was prepared by incorporating N,N'-(4,4'-ethylene biphenyl) bis(3-methoxy salicylidine imine) as ionophore in the plasticized PVC membrane containing bis(2-ethylhexyl) sebacate (DOS) as plasticizer. This proposed membrane works on the basis of a cation-exchange mechanism and shows a significant absorbance signal change on exposure to acetate buffer solution of pH 4.0 containing copper ion. The proposed sensing film displays a linear range of 0.01-32.0 μg mL(-1) with a limit of detection 0.008 μg mL(-1). Moreover, upon the introduction of a negatively charged lipophilic additive (oleic acid) into the membrane, the optode displayed enhanced sensitivity. In addition, satisfactory analytical sensing characteristics for determining copper (II) ion were obtained in terms of the selectivity, stability and reproducibility. The response time of the optode was less than 3 min, depending on the concentration of Cu(ІІ) ions. The optode membrane has been applied to determine Cu(II) in various real samples.
Product Description
Supelco
Bis(2-ethylhexyl) sebacate, Selectophore, ≥97.0%
Sigma-Aldrich
Bis(2-ethylhexyl) sebacate, ≥97.0% (GC)
Sigma-Aldrich
Bis(2-ethylhexyl) sebacate, technical grade, 90%
Sigma-Aldrich
Poly(vinyl chloride), average Mw ~43,000, average Mn ~22,000
Sigma-Aldrich
Poly(vinyl chloride), average Mw ~62,000, average Mn ~35,000
Sigma-Aldrich
Poly(vinyl chloride), average Mw ~233,000, average Mn ~99,000
Supelco
Poly(vinyl chloride), Selectophore, high molecular weight
Sigma-Aldrich
Poly(vinyl chloride), low molecular weight
Sigma-Aldrich
Poly(vinyl chloride), high molecular weight
Poly(vinyl chloride), European Pharmacopoeia (EP) Reference Standard
Supelco
Poly(vinyl chloride), analytical standard, average Mw 85,000 (Typical), average Mn 40,000 (Typical)