Merck
  • Associations between ambient air pollution and Hypertensive Disorders of Pregnancy.
Associations between ambient air pollution and Hypertensive Disorders of Pregnancy.
Environmental research (2013-03-26)
Zahra Mobasher, Muhammad T Salam, T Murphy Goodwin, Frederick Lurmann, Sue A Ingles, Melissa L Wilson
ABSTRACT
Exposure to ambient air pollution is linked to adverse pregnancy outcomes. Previous reports examining the relationship between ambient air pollution and Hypertensive Disorders of Pregnancy have been inconsistent. We evaluated the effects of ambient air pollution on the odds of Hypertensive Disorder of Pregnancy and whether these associations varied by body mass index (BMI). We conducted a retrospective, case-control study among 298 predominantly Hispanic women (136 clinically confirmed cases) who attended the Los Angeles County+University of Southern California Women's and Children's Hospital during 1996-2008. Trimester-specific carbon monoxide (CO), nitrogen dioxide (NO2), ozone (O3), and particulate matter with aerodynamic diameter <10 μm and <2.5 μm (PM10, PM2.5) exposure were estimated based on 24-hour exposure level at residential address. Logistic regression models were fitted to estimate odds ratios (ORs) and 95% confidence intervals (CIs) for two standard deviation increase in exposure levels. Exposures to CO and PM2.5 in the 1st trimester were significantly associated with Hypertensive Disorders of Pregnancy, and these associations were modified by BMI. In non-obese women (BMI <30), 1st trimester exposures to PM2.5 and CO were significantly associated with increased odds of Hypertensive Disorder of Pregnancy (ORs per 2-standard deviation increase in PM2.5 (7 μg/m(3)) and CO (1 ppm) exposures were 9.10 [95% CI: 3.33-24.6] and 4.96 [95% CI: 1.85-13.31], respectively). Additionally, there was a significantly positive association between exposure to O3 in the 2nd trimester and Hypertensive Disorder of Pregnancy (OR per 15 ppb=2.05; 95% CI: 1.22-3.46). Among non-obese women, 1st trimester exposure to PM2.5 and carbon monoxide are associated with increased odds of Hypertensive Disorder of Pregnancy.
Sigma-Aldrich
Nitrogen dioxide, ≥99.5%