Merck
  • Nocardiamides A and B, two cyclohexapeptides from the marine-derived actinomycete Nocardiopsis sp. CNX037.
Nocardiamides A and B, two cyclohexapeptides from the marine-derived actinomycete Nocardiopsis sp. CNX037.
Journal of natural products (2013-04-17)
Zheng-Chao Wu, Sumei Li, Sang-Jip Nam, Zhong Liu, Changsheng Zhang
ABSTRACT
Two new cyclic hexapeptides, nocardiamides A (1) and B (2), were isolated from the culture broth of marine-derived actinomycete CNX037 strain that was identified as a Nocardiopsis species. The planar structures of nocardiamides A (1) and B (2) were assigned on the basis of 1D and 2D NMR and HRESIMS spectroscopic analyses. Their absolute configurations were deduced by the advanced Marfey's method and chiral-phase HPLC analysis. The challenge of locating two d- and one l-valine residue in 1 and 2 was accomplished by total synthesis using solid-phase peptide synthetic methods. Both 1 and 2 showed negligible antimicrobial activities against seven indicator strains and exhibited no cytotoxicity against HCT-116.
Product Description
Supelco
L-Valine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SAFC
L-Valine
Sigma-Aldrich
L-Valine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Valine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Valine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Supelco
L-Valine, Pharmaceutical Secondary Standard; Certified Reference Material