Merck
Fluorimetric assay of acid lipase in human leukocytes.
Clinica chimica acta; international journal of clinical chemistry (1979-10-01)
S Kelly, R Bakhru-Kishore
PMID487606
ABSTRACT
A fluorimetric assay for leukocyte acid lipase, the enzyme defect in Wolman disease, is described. The method requires a smaller volume of blood and can be performed more rapidly than the present colorimetric assay for the leukocyte enzyme.