Merck
  • Inhibition by diacylmethane derivatives of mutagenicity and nucleic acid binding of 2-aminofluorene derivatives.
Inhibition by diacylmethane derivatives of mutagenicity and nucleic acid binding of 2-aminofluorene derivatives.
Journal of the National Cancer Institute (1989-11-15)
C Y Wang, M S Lee, H Nagase, K Zukowski
ABSTRACT
The active methylene compounds acetylacetone, 1,1,1-trifluoroacetylacetone, benzoylacetone, dibenzoylmethane, and 1,3-indandione inhibited the mutagenicity of 2-nitrofluorene in Salmonella typhimurium. They also inhibited the N,O-acetyltransferase-catalyzed transfer RNA binding of N-hydroxy-2-acetylaminofluorene, but they did not inhibit N,O-acetyltransferase. However, only 1,3-indandione and 1,1,1-trifluoroacetylacetone significantly inhibited the binding of N-acetoxy-2-acetylaminofluorene to transfer RNA. Reaction of the trifluoro compound with the acetoxy compound yielded 1-(N-2-fluorenylacetamido)acetone. These results demonstrate that active methylene compounds can inhibit mutagenicity and nucleic acid binding of chemical carcinogens.
Sigma-Aldrich
1,1,1-Trifluoro-2,4-pentanedione, 98%
Sigma-Aldrich
1,3-Indandione, 97%
Sigma-Aldrich
1-Phenyl-1,3-butanedione, 99%