Merck
  • A non-invasive technique for continuous monitoring of left ventricular function using a new solid state mercuric iodide radiation detector.
A non-invasive technique for continuous monitoring of left ventricular function using a new solid state mercuric iodide radiation detector.
Clinical science (London, England : 1979) (1984-05-01)
A Lahiri, J C Crawley, R I Jones, M J Bowles, E B Raftery
ABSTRACT
A miniature solid-state mercuric iodide (HgI2) nuclear probe detector has been developed in conjunction with a computerized nuclear probe (Nuclear Stethoscope) to enable continuous non-invasive monitoring of left ventricular function using 99Tc-labelled equilibrium blood pool techniques. Left ventricular ejection fraction was measured in 54 patients undergoing radionuclide angiography with a gamma-camera and with the Nuclear Stethoscope and a good correlation was obtained between both techniques (r = 0.94, n = 54, P less than 0.001). The prototype mercuric iodide detector was compared with the sodium iodide detector of the Nuclear Stethoscope and a study in 41 consecutive patients demonstrated a good correlation for the measurement of ejection fraction, ejection rate, peak filling rate and time to peak filling rate (r = 0.94, 0.89, 0.90 and 0.78 respectively). It may be possible to adapt the mercuric iodide detector for continuous non-invasive monitoring of left ventricular performance in critically ill patients and during physiological or pharmacological interventions.
Sigma-Aldrich
Mercury(II) iodide, ACS reagent, ≥99.0%
Sigma-Aldrich
Mercury(II) iodide red, puriss. p.a., ACS reagent, red, ≥99.0% (RT)