Merck
  • Prevention of Eimeria alabamensis coccidiosis by a long-acting baquiloprim/sulphadimidine bolus.
Prevention of Eimeria alabamensis coccidiosis by a long-acting baquiloprim/sulphadimidine bolus.
Veterinary parasitology (1998-04-30)
C Svensson
ABSTRACT
Twelve calves aged 6-10 months, and 12 calves aged 10-16 months were turned out onto a permanent pasture known to have been contaminated with oocysts of Eimeria alabamensis during the previous year. Two days after turnout, six of the older calves and six of the younger were each treated with one bolus per 200 kg bodyweight containing 1.6 g baquiloprim and 14.4 g sulphadimidine. The other 12 calves were left untreated. The excretion of Eimeria oocysts, the faecal dry matter and the weight gain of treated and untreated calves within each age group were compared during the first 3 weeks on pasture to assess the efficacy of the bolus in preventing E. alabamensis coccidiosis. All the older of the untreated calves and four of the younger developed gruel-like to watery diarrhoea 4-7 days after turnout. The faecal consistency of the treated calves remained firm and they lost significantly less weight than the control calves during the first 13 days on pasture. The treated calves also excreted significantly fewer oocysts during the first 20 days of grazing; their oocyst excretion remained low during days 8-10 when all but one of the diarrhoeic control calves excreted more than 850,000 oocysts per gram faeces (OPG). Starting on days 12 to 14 the oocyst excretion of 8 of the treated calves increased to 20,000-65,000 OPG and of 2 calves to 210,000-240,000 OPG. There was no difference in oocyst output between treated and untreated calves from the fourth week of grazing and no difference in weight gain among the younger calves. In the older calves there was a tendency for the untreated calves to gain more weight than treated calves.
Product Description
Supelco
Baquiloprim, VETRANAL®, analytical standard