Merck
  • The conversion of a phenol to an aniline occurs in the biochemical formation of the 1-(4-aminophenyl)-1-deoxy-D-ribitol moiety in methanopterin.
The conversion of a phenol to an aniline occurs in the biochemical formation of the 1-(4-aminophenyl)-1-deoxy-D-ribitol moiety in methanopterin.
Biochemistry (2011-06-04)
Robert H White
ABSTRACT
Recent work has demonstrated that 4-hydroxybenzoic acid is the in vivo precursor to the 1-(4-aminophenyl)-1-deoxy-D-ribitol (APDR) moiety present in the C(1) carrier coenzyme methanopterin present in the methanogenic archaea. For this transformation to occur, the hydroxyl group of the 4-hydroxybenzoic acid must be replaced with an amino group at some point in the biosynthetic pathway. Using stable isotopically labeled precursors and liquid chromatography with electrospray-ionization mass spectroscopy, the first step of this transformation in Methanocaldococcus jannaschii occurs by the reaction of 4-hydroxybenzoic acid with phosphoribosyl pyrophosphate (PRPP) to form 4-(β-d-ribofuranosyl)hydroxybenzene 5'-phosphate (β-RAH-P). The β-RAH-P then condenses with l-aspartate in the presence of ATP to form 4-(β-d-ribofuranosyl)-N-succinylaminobenzene 5'-phosphate (β-RFSA-P). Elimination of fumarate from β-RFSA-P produces 4-(β-D-ribofuranosyl)aminobenzene 5'-phosphate (β-RFA-P), the known precursor to the APDR moiety of methanopterin [White, R. H. (1996) Biochemistry 35, 3447-3456]. This work represents the first biochemical example of the conversion of a phenol to an aniline.
Sigma-Aldrich
Adonitol, ≥99%
Sigma-Aldrich
Adonitol, BioXtra, ≥99.0% (HPLC)