Merck
  • Influence of sildenafil on central dopamine-mediated behaviour in male rats.
Influence of sildenafil on central dopamine-mediated behaviour in male rats.
Life sciences (2002-03-16)
F Ferrari, A Ottani, D Giuliani
ABSTRACT
Two experiments were performed to evaluate the effects of sildenafil on spontaneous or dopamine agonist-induced behaviour in male rats. Data obtained in experiment 1 show that oral administration of the drug, at 1 mg/kg, significantly increased the occurrence of penile erections, anogenital self-grooming and homosexual mounting in grouped sexually-experienced, but not inexperienced, animals. In experiment 2, pre-treatment with sildenafil (0.5, 1 or 10 mg/kg) dose-dependently modified several behavioural signs, centrally evoked by the D2/D3 dopamine agonists, 7-OH-DPAT or B-HT 920 (both at 0.1 mg/kg), in experimentally naive male rats. While sildenafil at 1 mg/kg significantly increased the number of penile erection and stretching-yawning episodes induced by 7-OH-DPAT or B-HT 920, at 10 mg/kg it elicited low stereotyped behaviour, antagonizing stretching-yawning and sedation in 7-OH-DPAT treated rats. Discussion centres on the modulatory activity of sildenafil on central dopaminergic pathways and, possibly, on nitric oxide production.