- [Recommendation for using granulocyte and granulocyte-macrophage growth factor during anti-cancer therapy in children].
[Recommendation for using granulocyte and granulocyte-macrophage growth factor during anti-cancer therapy in children].
Przeglad lekarski (2006-08-09)
Walentyna Balwierz, Angelina Moryl-Bujakowska, Adriana Balwierz, Katarzyna Pawińska
PMID16892890
ABSTRACT
Neutropenia is the most often side effect during antineoplastic treatment. In this article current recommendations for clinical applications of granulocyte and granulocyto-macrophage hematopoietic factors and possibility of protection of neutropenia, and its serious complications in pediatric oncology are presented.