Merck
  • The reinforcing actions of a serotonin-3 receptor agonist within the ventral tegmental area: evidence for subregional and genetic differences and involvement of dopamine neurons.
The reinforcing actions of a serotonin-3 receptor agonist within the ventral tegmental area: evidence for subregional and genetic differences and involvement of dopamine neurons.
The Journal of pharmacology and experimental therapeutics (2007-02-28)
Zachary A Rodd, Victoria E Gryszowka, Jamie E Toalston, Scott M Oster, Dong Ji, Richard L Bell, William J McBride
ABSTRACT
Studies from our laboratory indicated that local perfusion of the ventral tegmental area (VTA) with a serotonin-3 (5-HT(3)) receptor agonist increased dopamine (DA) neuronal activity and that the self-infusion of ethanol (EtOH) and cocaine into the posterior VTA could be inhibited with coadministration of a 5-HT(3) receptor antagonist. The study tested the hypothesis that activating 5-HT(3) receptors within the VTA produces reinforcing effects. The study also examined whether there were differences between Wistar rats and a line of rats selectively bred for high alcohol consumption with regard to the self-infusion of a 5-HT(3) receptor agonist within the VTA. Adult female alcohol-preferring (P) and Wistar rats were allowed to self-infuse the 5-HT(3) receptor agonist 1-(m-chlorophenyl)-biguanide (CPBG) into the posterior or anterior VTA. Furthermore, experiments examined the effects of coinfusion of the 5-HT(3) antagonist ICS 205,930 (ICS), and the DA D(2,3) agonist quinpirole on the self-infusion of CPBG. Both Wistar and P rats readily self-administered CPBG into the posterior, but not anterior, VTA. P rats self-infused lower concentrations of CPBG (0.10 microM) than did Wistar rats (1.0 microM). Coinfusion of either ICS or quinpirole reduced CPBG self-infusion into the posterior VTA. The results of this study suggest that activation of 5-HT(3) receptors within the posterior VTA produces reinforcing effects and that these reinforcing effects are mediated through activation of DA neurons. Furthermore, the data suggest that selective breeding for alcohol-preference results in the posterior VTA being more sensitive to the reinforcing effects of CPBG.